Order Entry
Belgium
ContactUsLinkComponent
Papanicolaou's solution 3c Polychromatic solution EA 65 used in nuclear staining, for cytology, Sigma-Aldrich®
Flammable
Irritant
Papanicolaou's solution 3c Polychromatic solution EA 65 used in nuclear staining, for cytology, Sigma-Aldrich®
Catalog # 1.09270.0100
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Papanicolaou's solution 3c Polychromatic solution EA 65 used in nuclear staining, for cytology, Sigma-Aldrich®
Catalog # 1.09270.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Pk Info:
    300 - 400 stainings/100 ml

Specifications

Specification Test Results

  • Suitability for microscopy (Vaginal smear):
    passes test
  • Nuclei:
    blue to dark violet
  • Cytoplasm:
    red

Specification Test Results

About this item

 :
Density:0.82 g/cm3 (20 °C)
Flash Pt:15 °C
UN:1170
ADR:3,II
 :GHS02
 :GHS07
 :1