ContactUsLinkComponent
Papanicolaou's solution 1b Hematoxylin solution S used in nuclear staining, for cytology, Sigma-Aldrich®
Health
Irritant
Papanicolaou's solution 1b Hematoxylin solution S used in nuclear staining, for cytology, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Health
Irritant
Papanicolaou's solution 1b Hematoxylin solution S used in nuclear staining, for cytology, Sigma-Aldrich®
Supplier:  Merck
Certificates
Specification Test Results
  • Suitability for microscopy (Vaginal smear):
    passes test
  • Nuclei:
    blue to dark violet
  • Cyanophilic cytoplasm:
    blue-green
  • Eosinophilic cytoplasm:
    pink