Orders LinkContactUsLinkComponent
Kettle brushes
Kettle brushes
Supplier:   MICRONOVA
Kettle brushes
Supplier:  MICRONOVA
Certificates
Kettle brushes
3 Options Available Below
Product Details & Documents
The kettle brush is an oblong-shaped brush with a flare head design and polyester bristles, that is well suited for an all-around, thorough cleaning of small tanks, carboys and other production equipment.
  • Lightweight, durable brush with optional fabric covers
  • Made from FDA approved, BPA-free base resins
  • FDA approved for indirect food contact
  • Features three ACME threaded handle attachments, one at 90° and two angled on either side
  • Optional covers and handles sold separately
  • Autoclavable, with base down
Attaches to Micronova's STA series, fixed and extendible length handles to reach the bottom of large process tanks and kettles.
Product Family Options
Product Information
  • Bristle material
    Brush head length
    Brush head width
    Handle material
    Availability
    Price
  • MICRBRWH-41
    Kettle brush, small
    Kettle brush, small
    Polyester
    190 mm
    133 mm
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • MICRBRWH-42
    Kettle brush, large
    Kettle brush, large
    Polyester
    267 mm
    152 mm
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 194,00 €
    194,00 €
    1 * 1 ST
       194,00 €
  • MICRBRDB-41
    Kettle brush, small
    Kettle brush, small
    Polyester
    190 mm
    133 mm
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.