Supelco® Organic Reference Standard, Zingerone
Catalog # 88787-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:122-48-5
- Description:Zingerone
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00048232
- M-Clarity quality level:MQ100
- Synonym(s):Vanillylacetone; Zingerone
- Melting point:40 - 41 °C (lit.)
- Formula:4-(HO)C₆H₃-3-(OCH₃)CH₂CH₂COCH₃
- Molecular weight:194,23
- Density:1,14 g/ml at 25 °C (lit.)
- Flash point:113 °C - Closed cup
- Boiling point:141 °C/0,5 mmHg (lit.)