Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Zearalenone solution, analytical standard, Supelco®
Organic reference standard, Zearalenone solution, analytical standard, Supelco®
Catalog # 34126-2ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Zearalenone solution, analytical standard, Supelco®
Catalog # 34126-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
     17924-92-4
  • Description:
    Zearalenone solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₈H₂₂O₅
  • Storage temperature:
    −20 °C
  • Description:
    Zearalenone solution

Specifications

About this item