Organic reference standard, Xanthydrol solution, for the detection of urea, Supelco®
Catalog # 95582-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:~10% in methanol
- Pk:100 mL
- CAS No.:90-46-0
- Description:Xanthydrol solution
- Formula:C₁₃H₁₀O₂
- Storage temperature:2...8 °C
- Description:Xanthydrol solution