Supelco® Organic Reference Standard, Withaferin A
Catalog # 89910-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:5119-48-2
- Description:Withaferin A
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD10687098
- M-Clarity quality level:MQ100
- Synonym(s):4β,5β,6β,22R)-5,6-Epoxy-4,22,27-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic acid δ-lactone; Withaferine
- Molecular weight:470,60
- Flash point:226 °C