Supelco® Organic Reference Standard, Vitexin
49513-10MG-F
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:3681-93-4
- Description:Vitexin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00017456
- M-Clarity quality level:MQ100
- Synonym(s):4′,5,7-Trihydroxyflavone 8-C-glucoside; 8-C-Glucosylapigenin; Apigenin 8-C-glucoside; Orientoside
- Molecular weight:432,38