Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Trans-Cinnamaldehyde
Supelco® Organic Reference Standard, Trans-Cinnamaldehyde
Catalog # 06536-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Trans-Cinnamaldehyde
Catalog # 06536-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    14371-10-9
  • Description:
    trans-Cinnamaldehyde
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H317 - H319 - H335
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00007000
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    trans-3-Phenyl-2-propenal
  • Melting point:
    –9…+4 °C (lit.)
  • Formula:
    C₆H₅CH=CHCHO
  • Molecular weight:
    132,16
  • Storage temperature:
    2...8 °C
  • Density:
    1,05 g/ml at 25 °C (lit.)
  • Flash point:
    71 °C - closed cup
  • Boiling point:
    250...252 °C (lit.)

Specifications

About this item

  • GHS signal: GHS07
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338