Supelco® Organic Reference Standard, Sucrose
S1174-500MG
: Merck
:
- Pk:500 mg
- Standards:Analytical standard, for enzymatic assay kit SCA20
- CAS No.:57-50-1
- Description:Sucrose
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00006626
- M-Clarity quality level:MQ100
- Synonym(s):α-ᴅ-Glc-(1→2)-β-ᴅ-Fru; α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside; β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside; ᴅ(+)-Saccharose; Sugar
- Melting point:185 - 187 °C (lit.)
- Molecular weight:342,30