Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Senecionine, analytical standard, Supelco®
Organic reference standard, Senecionine, analytical standard, Supelco®
Catalog # 50351-5MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Senecionine, analytical standard, Supelco®
Catalog # 50351-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    130-01-8
  • Description:
    Senecionine
  • Formula:
    C₁₈H₂₅NO₅
  • Storage temperature:
    2 - 8 °C
  • Description:
    Senecionine
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.