Order Entry
Belgium
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Scutellarin
Catalog # 73577-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Scutellarin
Catalog # 73577-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    27740-01-8
  • Description:
    Scutellarin
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01861503
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4′,5,6,7-Tetrahydroxyflavone 7-glucuronide; 5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-1-benzopyran-7-yl-β-ᴅ-glucopyranosiduronic acid; 7-(β-ᴅ-Glucopyranuronosyloxy)-5,6-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one; Breviscapin; Scutellarein-7-O-β-ᴅ-glucuronide
  • Molecular weight:
    462,36

Specifications

About this item