Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Rutin trihydrate
Supelco® Organic Reference Standard, Rutin trihydrate
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Rutin trihydrate
Supplier:  Merck

Specifications

  • CAS No.:
    250249-75-3
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00149490
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Quercetin-3-rutinoside trihydrate; Vitamin P trihydrate
  • Molecular weight:
    664,56

Specifications

Frequently Bought Together