Supelco® Organic Reference Standard, Punicalagin
50793-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:65995-63-3
- Description:Punicalagin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD09838017
- M-Clarity quality level:MQ100
- Molecular weight:1084,72