Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Paraben internal standard mix solution, analytical standard, Supelco®
Organic reference standard, Paraben internal standard mix solution, analytical standard, Supelco®
Catalog # 32124-5ML
Supplier:  Merck
CAS Number:  
Organic reference standard, Paraben internal standard mix solution, analytical standard, Supelco®
Catalog # 32124-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetone
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • Description:
    Paraben internal standard mix solution
  • Storage temperature:
    –20 °C
  • Description:
    Paraben internal standard mix solution
  • Flash point:
    –17 °C

Specifications

About this item