Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Ochratoxin A-13C20 solution, analytical standard, Supelco®
Organic reference standard, Ochratoxin A-13C20 solution, analytical standard, Supelco®
Catalog # 33416-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Ochratoxin A-13C20 solution, analytical standard, Supelco®
Catalog # 33416-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    911392-42-2
  • Description:
    Ochratoxin A-13C20 solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    ₁₃C₂₀H₁₈ClNO₆
  • Storage temperature:
    –20 °C
  • Description:
    Ochratoxin A-13C20 solution

Specifications

About this item

Ochratoxin A is a mycotoxin, which is commonly produced by fungi Aspergillus ochraceus and Penicillium verrucosum in improperly stored food products of plant origin, edible animal tissues, human blood sera and tissues.