Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Loganin
Supelco® Organic Reference Standard, Loganin
Catalog # 36483-10MG-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Loganin
Catalog # 36483-10MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    18524-94-2
  • Description:
    Loganin
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00075645
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    7-Hydroxy 6-desoxyverbenalin; Loganoside
  • Formula:
    C₁₇H₂₆O₁₀
  • Molecular weight:
    390,38
  • Flash point:
    Not applicable
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.