Order Entry
Belgium
ContactUsLinkComponent
Phytochemical standard, Isosilybin A, analytical standard, Supelco®
Phytochemical standard, Isosilybin A, analytical standard, Supelco®
Catalogus # 97326-5MG
Leverancier:  Merck
CAS-nummer:  
undefined
Phytochemical standard, Isosilybin A, analytical standard, Supelco®
Catalogus # 97326-5MG
Leverancier:  Merck
CAS-nummer:  

Specificaties

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    142796-21-2
  • Description:
    Isosilybin A, ≥95,0% (HPLC)
  • Formula:
    C₂₅H₂₂O₁₀
  • Description:
    Isosilybin A
  • Form:
    Neat

Specificaties

Over dit artikel

Verpakking: Bottomless glass bottle. Contents are inside inserted fused cone.