Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, HPLC Protein Standard Mixture
Supelco® Organic Reference Standard, HPLC Protein Standard Mixture
Catalog # H2899-1VL
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, HPLC Protein Standard Mixture
Catalog # H2899-1VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    HPLC protein standard mixture
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H334
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ200

Specifications

About this item

Caution: P261 - P284 - P501