Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Genistin
Supelco® Organic Reference Standard, Genistin
  73822-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Genistin
  73822-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    529-59-9
  • Description:
    Genistin
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00016883
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4′,5,7-Trihydroxyisoflavone 7-glucoside; Genistein 7-glucoside; Genistein-7-O-β-ᴅ-glucopyranoside; Genistoside; NSC 5112
  • Molecular weight:
    432,38