Organic reference standard, ethyl pyruvate, analytical standard, Supelco®
Catalog # 42327-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:617-35-6
- Description:Ethyl pyruvate, ≥97,0% (GC)
- Formula:C₅H₈O₃
- Storage temperature:2...8 °C
- Description:Ethyl pyruvate
- Boiling point:144 °C (lit.)