Organic reference standard, Dibenzyl ether, analytical standard, Supelco®
: Merck
- CAS No.:103-50-4
- Melting point:1,5...3,5 °C (lit.)
- Formula:C₁₄H₁₄O
- Description:Dibenzyl ether
- Flash point:135 °C
- Boiling point:158...160 °C/10 mmHg (lit.)
Frequently Bought Together