Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, δ-Tocotrienol
Supelco® Organic Reference Standard, δ-Tocotrienol
Catalog # 69745-100MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, δ-Tocotrienol
Catalog # 69745-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    25612-59-3
  • Description:
    δ-Tocotrienol
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD11045309
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (R)-δ-Tocotrienol; 8-Methyltocotrienol; [R-(E,E)]-3,4-Dihydro-2,8-dimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol
  • Molecular weight:
    396,61

Specifications

About this item