Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Antioxidant Kit 2, Phenolics, analytical standard, Supelco®
Organic reference standard, Antioxidant Kit 2, Phenolics, analytical standard, Supelco®
Catalog # 40048-U
Supplier:  Merck
CAS Number:  
Organic reference standard, Antioxidant Kit 2, Phenolics, analytical standard, Supelco®
Catalog # 40048-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Individually packaged in quantities indicated
  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    Antioxidant Kit 2
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    RT
  • Description:
    Antioxidant Kit 2
  • Pk Info:
    1 Kit
  • Form:
    Neat

Specifications

About this item