Organic reference standard, 4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard, Supelco®
Catalog # 90034-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:1135-24-6
- Description:4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, ≥99,0% (HPLC)
- Formula:C₁₀H₁₀O₄
- Storage temperature:2...8 °C
- Description:4-Hydroxy-3-methoxycinnamic acid
- Form:Neat