Order Entry
Belgium
ContactUsLinkComponent
 
Organic reference standard, 3-Octanol, analytical standard, Supelco®
Catalog # 93856-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 3-Octanol, analytical standard, Supelco®
Catalog # 93856-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    589-98-0
  • Description:
    3-Octanol, ≥98,5% (GC)
  • Formula:
    C₈H₁₈O
  • Description:
    3-Octanol
  • Flash point:
    68 °C
  • Boiling point:
    174...176 °C (lit.)
  • Form:
    Neat

Specifications

About this item