Order Entry
Belgium
Orders LinkContactUsLinkComponent
Organic reference standard, 2-Methylcyclohexanone, analytical standard, Supelco®

Organic reference standard, 2-Methylcyclohexanone, analytical standard, Supelco®

2-Methylcyclohexanone, ≥98,0% (GC), Analytical standard, 1 mL
Catalog # 44479-1ML
Supplier:   Merck
Organic reference standard, 2-Methylcyclohexanone, analytical standard, Supelco®
Catalog # 44479-1ML
Supplier:  Merck
CAS Number:  
Organic reference standard, 2-Methylcyclohexanone, analytical standard, Supelco®
Organic reference standard, 2-Methylcyclohexanone, analytical standard, Supelco®

2-Methylcyclohexanone, ≥98,0% (GC), Analytical standard, 1 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • Description:
    2-Methylcyclohexanone, ≥98,0% (GC)
  • Cat. No.:
    44479-1ML
  • Formula:
    C₇H₁₂O
  • Description:
    2-Methylcyclohexanone
  • Boiling point:
    162...163 °C (lit.)
  • CAS No.:
    583-60-8

Specifications