Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, 2-Methoxy-4-methylphenol, analytical standard, Supelco®
Organic reference standard, 2-Methoxy-4-methylphenol, analytical standard, Supelco®
  41340-1ML
 :  Merck
 :  
undefined
Organic reference standard, 2-Methoxy-4-methylphenol, analytical standard, Supelco®
  41340-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    93-51-6
  • Description:
    2-Methoxy-4-methylphenol, ≥98,0% (GC)
  • Melting point:
    5 °C (lit.)
  • Formula:
    C₈H₁₀O₂
  • Description:
    2-Methoxy-4-methylphenol
  • Boiling point:
    221...222 °C (lit.)