Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, [8]-Gingerol
Supelco® Organic Reference Standard, [8]-Gingerol
Catalog # 01514-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, [8]-Gingerol
Catalog # 01514-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    23513-08-8
  • Description:
    [8]-Gingerol
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00877794
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-dodecanone; 8-Gingerol
  • Formula:
    C₁₉H₃₀O₄
  • Molecular weight:
    322,44
  • Storage temperature:
    2…8 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338