Order Entry
Belgium
Orders LinkContactUsLinkComponent
Squeegees, Plastic

Squeegees, Plastic

Supplier:   MICRONOVA
Squeegees, Plastic
Supplier:  MICRONOVA
Certificates
Squeegees, Plastic
Squeegees, Plastic

5 Options Available Below

Product Details & Documents

These plastic, double foam squeegees are easy to sanitise.

  • Will not rust
  • Extremely durable
  • Synthetic rubber handle with PP frame
  • For use with HACCP compliance programs and colour coding

Integrated blade eliminates seams and crevices, reducing places for bacteria to grow. Frame handle is threaded for use with STA series of plastic handles.

Product Family Options

Product Information

  • Colour
    Length
    Availability
    Price
  • MICRSQDG-24
    Plastic squeegee with foam blade
    Plastic squeegee with foam blade
    Green
    610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €

  • MICRSQRE-24
    Plastic squeegee with foam blade
    Plastic squeegee with foam blade
    Red
    610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €

  • MICRSQWH-24
    Plastic squeegee with foam blade
    Plastic squeegee with foam blade
    White
    610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €

  • MICRSQYE-24
    Plastic squeegee with foam blade
    Plastic squeegee with foam blade
    Yellow
    610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €

  • MICRSQWH-24R
    Plastic squeegee with rubber blade
    Plastic squeegee with rubber blade
    White
    610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.