Order Entry
Belgium
ContactUsLinkComponent
n-Paraffin Mix C24, C28, C32, C36, certified reference material, Supelco®
n-Paraffin Mix C24, C28, C32, C36, certified reference material, Supelco®
Catalog # 47107.
Supplier:  Merck
CAS Number:  
n-Paraffin Mix C24, C28, C32, C36, certified reference material, Supelco®
Catalog # 47107.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2 % (w/w) each component in octane
  • Pk:
    1
  • Description:
    n-Paraffin Mix C24, C28, C32, C36

Specifications

About this item