Order Entry
Belgium
ContactUsLinkComponent
ASTM® D5442 quantitative linearity standard, C16 - C44, analytical standard, Supelco®
ASTM® D5442 quantitative linearity standard, C16 - C44, analytical standard, Supelco®
Catalog # 502286.
Supplier:  Merck
CAS Number:  
ASTM® D5442 quantitative linearity standard, C16 - C44, analytical standard, Supelco®
Catalog # 502286.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.01 % (w/w) each component in cyclohexane
  • Pk:
    1
  • Description:
    ASTM(R) D5442 Quantitative Linearity Standard, C16 - C44

Specifications

About this item