Order Entry
Belgium
ContactUsLinkComponent
ASTM® D5441 MTBE contaminants (low) mix B, analytical standard, Supelco®
ASTM® D5441 MTBE contaminants (low) mix B, analytical standard, Supelco®
Catalog # 47943.
Supplier:  Merck
CAS Number:  
ASTM® D5441 MTBE contaminants (low) mix B, analytical standard, Supelco®
Catalog # 47943.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.1 % (w/w) each component in methyl tert-butyl ether
  • Pk:
    1
  • Description:
    ASTM(R) D5441 MTBE Contaminants (low) Mix B

Specifications

About this item