Order Entry
Belgium
ContactUsLinkComponent
ASTM® D4815 Qualitative peak ID mix, analytical standard, Supelco®
ASTM® D4815 Qualitative peak ID mix, analytical standard, Supelco®
Catalog # 47213.
Supplier:  Merck
CAS Number:  
ASTM® D4815 Qualitative peak ID mix, analytical standard, Supelco®
Catalog # 47213.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    (each component at the nominal wt% indicated)
  • Pk:
    1
  • Description:
    ASTM(R) D4815 Qualitative Peak ID Mix

Specifications

About this item