ContactUsLinkComponent
Control slides, TISSUE-TROL™, Sigma-Aldrich
Control slides, TISSUE-TROL™, Sigma-Aldrich
Catalog # TTR011-25EA
undefined
Control slides, TISSUE-TROL™, Sigma-Aldrich
Catalog # TTR011-25EA
Specifications
  • Description:
    Spirochetes TISSUE-TROL™ Control Slides from rabbit testicle tissue containing spirochetes (Treponema pallidum)
  • Pk:
    25
Specifications
About this item
Delivery: Box contains 24 unstained slides plus one reference slide.