ContactUsLinkComponent
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Catalog # EM0200-1KT
CAS Number:  
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Catalog # EM0200-1KT
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Osteo-Bed Bone Embedding Kit, Sigma-Aldrich®
Specifications
About this item
Suitable for use with large and small mineralized (undecalcified) bone sections. Yields clear, hard blocks for cutting sections. Not water-soluble.
  • Quality Level 200
Kit contains:
Osteo-Bed Bone Embedding Resin A (O8514) 900 ml
Osteo-Bed Bone Embedding Catalyst (O8764) 2 x 12 g