Order Entry
Belgium
ContactUsLinkComponent
N,N-Dimethylformamide dimethyl acetal methylation reagent, for GC derivatisation
Flammable
Irritant
N,N-Dimethylformamide dimethyl acetal methylation reagent, for GC derivatisation
Catalog # MANA701430.110
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Flammable
Irritant
N,N-Dimethylformamide dimethyl acetal methylation reagent, for GC derivatisation
Catalog # MANA701430.110
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pack type:
    Glass vial, amber, crimped
  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    1× 10 ml

Specifications

About this item

 :
 :119.16 g/mol
Boiling Pt:106–107 °C (760 mm Hg)
Density:0.897 g/cm³ (d20°/4°)
UN:3316
ADR:9,II
 :GHS02
 :GHS07
CAS Number:4637-24-5
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482