

N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, LiChropur™ silylation reagent, for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:24589-78-4
:
Formula:C₆H₁₂F₃NOSi
:199.25 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ100
UN:1993
:GHS02
:GHS07
Specification Test Results
- Appearance (Colour):Colourless
- Appearance (Form):Liquid
- Purity (GC area):≥97,0%
- GC minor component 1 (Area%):0,8 - 1,5 %
- Minor component 1:Trimethylchlorosilane
- Refractive index N20/D:1,377 - 1,380
- Proton NMR spectrum:Conforms to structure
Specification Test Results
Frequently Bought Together