Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (by GC), LiChropur™ for GC derivatisation, Supelco®
Flammable
Irritant
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (by GC), LiChropur™ for GC derivatisation, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (by GC), LiChropur™ for GC derivatisation, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Appearance (Colour):
    Colorless to Very Light Yellow
  • Appearance (Form):
    Liquid
  • Purity (GC area %):
    ≥ 90.0 %
  • Refractive index n20/D:
    1.351 - 1.355
  • Infrared spectrum:
    Conforms to structure

Specification Test Results

Related Information

Frequently Bought Together