Pesticide standard, Metosulam, reference material, Honeywell Fluka™
HONCR1689-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:139528-85-1
- Description:Metosulam
- Environmentally Preferable:
- Formula:C₁₄H₁₃Cl₂N₅O₄S
- Storage temperature:Ambient
- Description:Metosulam
- Form:Powder
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.