Pesticide standard, Triflumuron, reference material, Honeywell Fluka™
Catalog # HONCR1683-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:64628-44-0
- Description:Triflumuron
- Formula:C₁₅H₁₀ClF₃N₂O₃
- Storage temperature:Ambient
- Description:Triflumuron
- Form:Crystalline
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.