Pesticide standard, (E)-Metominostrobin, reference material, Honeywell Fluka™
HONCR1662-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:133408-50-1
- Description:(E)-Metominostrobin
- Formula:C₁₆H₁₆N₂O₃
- Storage temperature:Ambient
- Description:(E)-Metominostrobin
- Form:Crystalline powder
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.