Pesticide standard, Fenbuconazole, reference material, Honeywell Fluka™
HONCR1639-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:114369-43-6
- Description:Fenbuconazole
- Environmentally Preferable:
- Formula:C₁₉H₁₇ClN₄
- Storage temperature:Ambient
- Description:Fenbuconazole
- Form:Crystalline
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.