Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide activated I, LiChropur™ for GC derivatisation activated with ethanethiol and ammonium iodide, Supelco®
Flammable
Irritant
Environment
N-Methyl-N-(trimethylsilyl)trifluoroacetamide activated I, LiChropur™ for GC derivatisation activated with ethanethiol and ammonium iodide, Supelco®
Supplier:  Merck
Certificates
Flammable
Irritant
Environment
N-Methyl-N-(trimethylsilyl)trifluoroacetamide activated I, LiChropur™ for GC derivatisation activated with ethanethiol and ammonium iodide, Supelco®
Supplier:  Merck

Specification Test Results

  • Appearance (Colour):
    Colorless
  • Appearance (Form):
    Liquid
  • Remarks on GC:
    Suitable for the silylation of testosterone
  • Refractive index N20/D:
    1.379 - 1.381
  • Proton NMR spectrum:
    Confroms to structure

Specification Test Results

Frequently Bought Together