ContactUsLinkComponent
Magnetic stirring bar, octagonal, PTFE
Magnetic stirring bar, octagonal, PTFE
Catalog # COWIFT.8360
Supplier:  COWIE
undefined
Magnetic stirring bar, octagonal, PTFE
Catalog # COWIFT.8360
Supplier:  COWIE
Specifications
  • Ø:
    13 mm
  • Length:
    75 mm
  • Pk:
    1
Specifications
About this item
Octahedral magnetic stirring bars.
  • Octahedral PTFE Stirrer Bar with champhered magnet core
  • 100% visual inspection using 4× magnification
Delivery: Supplied with a free of charge certificate of analysis confirming the Job Number, material batch number, date of manufacture and inspection criteria.