Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
VOAs in soil 2 - medium level, Supelco®
VOAs in soil 2 - medium level, Supelco®
VOAs in soil 2 - medium level, Supelco®
Catalog # SPE051H-25G
Supplier:
Merck
CAS Number:
Expand image
VOAs in soil 2 - medium level, Supelco®
Catalog # SPE051H-25G
Supplier:
Merck
CAS Number:
Specifications
Pk:
25 g
Description:
VOAs in soil 2 - medium level
Specifications
About this item
Product Details
Product Details