Order Entry
Belgium
ContactUsLinkComponent
Pesticides 1 - WP, proficiency testing material, Supelco®
Pesticides 1 - WP, proficiency testing material, Supelco®
Catalog # PE1280-1.2ML
Supplier:  Merck
CAS Number:  
Pesticides 1 - WP, proficiency testing material, Supelco®
Catalog # PE1280-1.2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1 mL for dilution up to 1L
  • Pk:
    1,2 mL
  • Description:
    Pesticides 1 - WP
  • Type:
    Proficiency Testing Material

Specifications

About this item