Order Entry
Belgium
ContactUsLinkComponent
Chlordane (Total) - WP, proficiency testing material, Supelco®
Chlordane (Total) - WP, proficiency testing material, Supelco®
Catalog # PE1092-2ML
Supplier:  Merck
CAS Number:  
Chlordane (Total) - WP, proficiency testing material, Supelco®
Catalog # PE1092-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Concentrate for dilution to 1L
  • Pk:
    2 mL
  • Description:
    Chlordane (Total) - WP
  • Type:
    Proficiency Testing Material

Specifications

About this item