SS TCL Polynuclear Aromatic Hydrocarbons Mix, certified reference material, Supelco®
Catalog # 4S8905
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 mug/mL each component in methylene chloride: benzene (1:1)
- Pk:1
- Description:SS TCL Polynuclear Aromatic Hydrocarbons Mix
- Type:Certified reference material