EPA 505/525 update pesticides mix A, certified reference material, Supelco®
Catalog # 47727-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:500 mug/mL each component in acetone
- Pk:1
- Description:EPA 505/525 Update Pesticides Mix A
- M-Clarity quality level:MQ100
- Type:Certified reference material